Compact Glove Boxes, Plas-Labs™
Supplier:   Plas-Labs
Compact Glove Boxes, Plas-Labs™
Supplier:  Plas-Labs
Certificates
Compact Glove Boxes, Plas-Labs™
Compact Glove Boxes, Plas-Labs™
9 Options Available Below
Product Details & Documents
Designed specifically for use when weighing hazardous materials that must be contained, glove boxes provide cleanliness, safety, portability, and a draft-free atmosphere for the maximum effectiveness of your balance
Top section is removable for easy installation of equipment, such as analytical balances, top loaders, bench scales, and moisture analyzers. Basic features include four purging gas valves (two on the transfer chamber and two on the main chamber), a white leveling tray for easy transferring of liquids, white ambidextrous Hypalon® gloves for superior chemical and UV resistance, a pressure relief valve that minimizes glove "fight back," and replaceable HEPA and organic vapor canister filters. All clamps are stainless steel and completely adjustable. Transfer chamber inner dimensions: 30.5Lx27.9Dcm (12x11").
Hospital-grade multiple electrical outlet strip is UL listed and CSA certified. CE marked.
Beneficial when working with toxic substances, asbestos fibers, sewage residue, and harmful liquid vapors. Ideal for determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics, and light radioactive materials. Minimize product degradation caused by moisture absorption.
 : Supplied with one-year manufacturer's warranty and instruction manual. Custom sizes are available. Models with additional glove ports are available. Exterior dimensions include transfer chamber. Allow 10cm (4") for elbow valve opposite transfer chamber. Contact your Avantor sales representative for information.
Product Family Options
Product Information
  • Désignation
    Volume
    Exterior Dimensions
    Interior Dimensions
    Door Opening
    Shipping Weight
    Availability
    Price
  • Glove Box with Transfer Chamber
    498 L (17⅗ cu. ft.)
    125W × 77D × 79H cm (49 × 30 × 31")
    89W × 74D × 77H cm (35 × 29 × 30")
    38×45 cm (15×18")
    95.3 kg (210 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $70,974.40
    $70,974.40
    Each
       $70,974.40
    Supplier Number: 830-ABB/EXP
  • Glove Box with Transfer Chamber
    730 L (25⅘ cu. ft.)
    160W × 78D × 86H cm (63 × 31 × 34")
    122W × 73D × 81H cm (48 × 29 × 32")
    38×45 cm (15×18")
    143 kg (315 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $69,652.42
    $69,652.42
    Each
       $69,652.42
    Supplier Number: 830-ABD/EXP
  • Glove Box with Transfer Chamber
    317 L (11⅕ cu. ft.)
    110W × 61D × 79H cm (43 × 24 × 31")
    71W × 59D × 74H cm (28 × 23 × 29")
    38×45 cm (15×18")
    90 kg (200 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $62,630.01
    $62,630.01
    Each
       $62,630.01
    Supplier Number: 830-ABC/EXP
  • Glove Box with Access Door
    317 L (11⅕ cu. ft.)
    125W × 77D × 79H cm (49 × 30 × 31")
    89W × 74D × 77H cm (35 × 29 × 30")
    54W × 54H cm (21½ × 21½")
    77.1 kg (170 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $80,893.34
    $80,893.34
    Each
       $80,893.34
    Supplier Number: 830CP-10/EXP
  • Glove Box with Access Door and Vent Outlet
    317 L (11⅕ cu. ft.)
    160W × 78D × 86H cm (63 × 31 × 34")
    122W × 73D × 81H cm (48 × 29 × 32")
    54W × 61H cm (21½ × 24")
    77.1 kg (170 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $62,490.92
    $62,490.92
    Each
       $62,490.92
    Supplier Number: 830CP-11/EXP
  • Glove Box with Access Door
    498 L (17⅗ cu. ft.)
    125W × 77D × 79H cm (49 × 30 × 31")
    89W × 74D × 77H cm (35 × 29 × 30")
    54W × 54H cm (21½ × 21½")
    97.5 kg (215 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $61,899.65
    $61,899.65
    Each
       $61,899.65
    Supplier Number: 830CP-20/EXP
  • Glove Box with Access Door and Vent Outlet
    498 L (17⅗ cu. ft.)
    160W × 78D × 86H cm (63 × 31 × 34")
    122W × 73D × 81H cm (48 × 29 × 32")
    54W × 61H cm (21½ × 24")
    97.5 kg (215 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $63,956.55
    $63,956.55
    Each
       $63,956.55
    Supplier Number: 830CP-21/EXP
  • Glove Box with Access Door
    730 L (25⅘ cu. ft.)
    125W × 77D × 79H cm (49 × 30 × 31")
    89W × 74D × 77H cm (35 × 29 × 30")
    54W × 54H cm (21½ × 21½")
    158.8 kg (350 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $71,691.51
    $71,691.51
    Each
       $71,691.51
    Supplier Number: 830CP-30/EXP
  • Glove Box with Access Door and Vent Outlet
    730 L (25⅘ cu. ft.)
    160W × 78D × 86H cm (63 × 31 × 34")
    122W × 73D × 81H cm (48 × 29 × 32")
    54W × 61H cm (21½ × 24")
    158.8 kg (350 lbs.)
    Log in for availability
    Log in for availability
    Each
    Each - $76,729.46
    $76,729.46
    Each
       $76,729.46
    Supplier Number: 830CP-31/EXP