VWR® FID Gas Stations
Catalog # 26000-036
Supplier: VWR International
CSA Certified
UL Listed
Specifications
- Description:VWR® Dual Gas Generator FID-6
- Dimensions:26.7W x 43.2D x 41.9H cm (101/2 x 17 x 161/2")
- Cat. no.:26000-036
- Flow Capacity:2500 cc/min. (zero air); 250 cc/min. (hydrogen)
- Weight:24 kg (53 lbs.)
Specifications
About this item
VWR® FID Gas Stations are designed to provide fuel gas and support air to flame ionization detectors, flame photometric detectors, and total hydrocarbon analyzers.
- Eliminate dangerous gas cylinders from lab
- Recommended and used by major GC and column manufacturers
VWR® FID Gas Stations provide both UHP zero air at purity levels below 0.05ppm total hydrocarbon content and hydrogen gas at purity levels above 99.9995% in one enclosure. Zero air is produced by purifying on-site compressed air. Pure hydrogen gas is generated by the electrolytic dissociation of water before passing through a desiccant cartridge. Systems increase the accuracy of analysis and reduce the need to clean detectors.