VWR® UHPN₂ Nitrogen Generator
CA26000-008
- Description:VWR® UHPN₂ Nitrogen generator
- Max. outlet flow:1100 cc/min. at 125 psig inlet air pressure, 1000 cc/min. at 110 psig inlet air pressure, 900 cc/min. at 100 psig inlet air pressure, 800 cc/min. at 90 psig inlet air pressure, 700 cc/min. at 80 psig inlet air pressure, 600 cc/min. at 70 psiginlet air pressure, 500 cc/min. at 60 psig inlet air pressure
- Max. outlet pressure:85 psig at 125 psig inlet air pressure, 75 psig at 110 psig inlet air pressure, 65 psig at 100 psig inlet air pressure, 60 psig at 90 psig inlet air pressure, 50 psig at 80 psig inlet air pressure, 45 psig at 70 psig inlet air pressure, 35 psig at 60 psig inlet air pressure
- Dimensions:30.5W x 40.6D x 88.9H cm (12 x 16 x 35")
- Weight:50 kg (110 lbs.)
- Cat. no.:CA26000-008
These systems are engineered to transform standard compressed air into 99.99% or 99.9999% nitrogen.
- Eliminate dangerous gas cylinders from lab
- Use for Gas Chromatography, LC/MS, ICP, and Thermal Analysis
Nitrogen purification utilizes pressure swing adsorption for the removal of oxygen, carbon dioxide, and water vapor. After nitrogen is purified, the gas is filtered through high efficiency coalescing prefilters and a 0.01 µm (absolute) membrane filter. The generators are enclosed in small cabinets that fit on or beneath most benchtop work surfaces. Best for carrier gas applications.