Protector® Glove Box Base Stands, Labconco® | Avantor
Protector® Glove Box Base Stands, Labconco®
Base stand, mobile, for use with Protector Double Glove Boxes, 249 kg (550 lbs.)
Catalog # 15203-116
Supplier:   Labconco
Fournisseur #:  5062020
Protector® Glove Box Base Stands, Labconco®
Catalog # 15203-116
Supplier:  Labconco
Protector® Glove Box Base Stands, Labconco®
Protector® Glove Box Base Stands, Labconco®
Base stand, mobile, for use with Protector Double Glove Boxes, 249 kg (550 lbs.)
Price
$10,300.00
Log in pour la disponibilité
Each
Product Details & Documents
Epoxy-coated, welded steel base stands have a permanently attached, black phenolic laminate work surface
Stands for single glove boxes have a work surface measuring 152.4Wx78.7Dx2.8Hcm (60x31x11/8"). Height is adjustable from 83.8–101.6cm (33–40") high. Mobile stand 97000-032 features 12.7cm (5") swivel casters and locking brakes. Fixed stand 97000-034 has adjustable leveling feet.
Stand 15203-116 supports a double glove box. Work surface measures 264.1Wx78.7Dx2.8Hcm (104x31x11/8"). Height is adjustable from 83.8–101.6cm (33–40") high with swivel casters.
Lower shelf provides convenient space for an accessory vacuum pump or optional drying train.
Spécifications
  • Désignation:
    Base stand, mobile, for use with Protector Double Glove Boxes
  • Shipping Weight:
    249 kg (550 lbs.)