ContactUsLinkComponent
EasySep™ Human Th17 Cell Enrichment Kit II
Catalogue # MSPP-17862
Fournisseur:  STEMCELL Technologies
undefined
EasySep™ Human Th17 Cell Enrichment Kit II
Catalogue # MSPP-17862
Fournisseur:  STEMCELL Technologies
Supplier Number:  17862
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Capacity:
    2×10⁹ cells
  • Description:
    EasySep™ human Th17 cell enrichment kit II
  • Cat. no.:
    MSPP-17862
Spécifications
À propos de cet article
Immunomagnetic positive selection of human Th17 cells.
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
The EasySep™ Human Th17 Enrichment Kit II is a two-step method designed to enrich for human Th17 cells from fresh peripheral blood mononuclear cells. First, CCR6+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™. Then, bound magnetic particles are removed from the cells and unwanted non-CD4+ T cells, CD45RA+ cells, and CXCR3+ cells are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™. The final isolated fraction is enriched for IL-17-producing Th17 cells.
This product can be used in place of the EasySep™ Human Th17 Cell Enrichment Kit for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').