ContactUsLinkComponent
EasySep™ Human B Cell Enrichment Kit II Without CD43 Depletion
Catalogue # MSPP-17963
Fournisseur:  STEMCELL Technologies
undefined
EasySep™ Human B Cell Enrichment Kit II Without CD43 Depletion
Catalogue # MSPP-17963
Fournisseur:  STEMCELL Technologies
Supplier Number:  17963
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ human B cell enrichment kit II without CD43 depletion
  • Cat. no.:
    MSPP-17963
Spécifications
À propos de cet article
Immunomagnetic isolation of untouched human B cells without CD43 depletion.
  • Fast and easy-to-use
  • Up to 99% purity
  • Isolated cells are untouched
The EasySep™ Human B Cell Enrichment Kit II without CD43 Depletion is designed to isolate B cells by negative selection from fresh or previously frozen PBMCs of individuals with B cell leukemia or lymphoma, or other disease states in which B cells may express CD43. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This kit replaces the EasySep™ Human B Cell Enrichment Kit without CD43 for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').