To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ non-human primate CD8+ T cell isolation kit
- Cat. no.:MSPP-19583
Specifications
About this item
Immunomagnetic isolation of untouched non-human primate CD8+ T cells
- Fast, easy to use, and column-free
- Up to 85% purity with high recovery
- Untouched, viable cells
The EasySep™ Non-Human Primate CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This kit has been verified for use with rhesus and cynomolgus macaques.