To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Capacity:1×10⁹ cells
- Description:EasySep™ 'Do-It-Yourself' positive selection kit II
- Cat. no.:MSPP-17698
Spécifications
À propos de cet article
Immunomagnetic positive selection of any cells with your own mouse IgG1 antibody.
- Fast and easy-to-use
- No columns
The EasySep™ 'Do-It-Yourself' Selection Kit II is designed to select any cell type of interest with your own mouse IgG1 monoclonal antibody. Cells that express the surface antigen that is recognized by the mouse IgG1 monoclonal antibody are targeted for magnetic separation using EasySep™ Dextran RapidSpheres™ and the EasySep™ Magnet.
This product replaces the EasySep™ 'Do-It-Yourself' Selection Kit (STMC18098).