Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Capacity:1×10⁹ cells
- Description:EasySep™ human ILC2 enrichment kit
- Cat. no.:MSPP-17972
Spécifications
À propos de cet article
Immunomagnetic enrichment of untouched human ILC2 cells.
- Fast, easy-to-use and column-free
- Untouched, viable cells
- Facilitates rapid flow sorting of ILC2s
The EasySep™ Human ILC2 enrichment kit is designed to enrich Group 2 Innate Lymphoid Cells (ILC2s) from leukapheresis products by immunomagnetic negative selection. Unwanted cells are targeted for removal by Tetrameric Antibody Complexes directed against non-ILC2s and dextran-coated magnetic particles (RapidSpheres™). Labeled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube as desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry and cell sorting.