KT™ KT5+ Premium Advanced Adhesive Slides
Fournisseur: StatLab
New Product
Certificats
À propos de cet article
KT™ Slides KT5+ Premium German advanced adhesion slides are an excellent choice for H and E, special stains, frozen sections, and even the toughest IHC procedures with difficult tissues.
- PiSmart S2 slide printer validated and compatible with both thermal transfer and UV laser printers
- Colored marking areas for efficient specimen color-coding, identification, labeling, and printing
- Superior adhesion maintains tissue integrity throughout the toughest of tissues or protocols
- No background staining
- Hybrid waterbath behavior: the tissue section jumps to the slide but water remains underneath for repositioning if necessary
Available in nine unique colors, in both clipped and 90° corners. Cold room storage is recommended.
Packaging: Sold by the case. 144 slides per gross, 10 gross/case.
Frequently Bought Together