LabExact® 934-AH® Glass Microfiber Filter, Prewashed and Dried, Not Tared, TSS or VSS Procedure
Fournisseur: I.W. TREMONT CO., INC.
New Product
Certificats
À propos de cet article
LabExact® Pre-washed and dried Grade 934-AH® filters for Total Suspended Solids (TSS) and Volatile Suspended Solids (VSS) are processed using reagent grade water (RGW) under strict controls to wash away extraneous fiber per the requirements of the EPA Standard 2540 C, D and E.
- Prepared filters are an effective time saver that removes a step from your work flow while maintaining your high levels of quality assurance
- Filters are packaged in plastic container, in stacks without weighing pans or bar coding
- Available in both 47 and 90 mm diameters
- Package of 100 washed and ready to use TSS and or VSS Grade 934-AH® filters
LabExact® Pre-washed and dried Grade 934-AH® filters for Total Suspended Solids (TSS) and Volatile Suspended Solids (VSS) are processed using reagent grade water (RGW) under strict controls to wash away extraneous fiber per the requirements of the EPA Standard 2540 C, D and E.
The trademark 934-AH® is registered and owned by Whatman Inc and used under license.
Packaging: Box of 100 filters.
Frequently Bought Together