LabExact® Grade TSS Glass Fiber Filter, Prewashed, Dried, Preweighed for Standard Methods TSS
Fournisseur: I.W. TREMONT CO., INC.
New Product
Certificats
À propos de cet article
LabExact® Total Suspended Solids (TSS) filters are processed to US EPA Standard Method 2540 C, D and E, washed using reagent grade water, dried, desiccated and weighed to 4 decimal place resolution.
- Each TSS binderless glass microfiber filter with 1.5 µm retention is individually packaged in a pan with a human readable serial number, weight and barcode for scanning
- Available in both 47 and 90 mm diameters
- Package of 100 washed, dried and tared (TSS) filters and pans
LabExact® Total Suspended Solids (TSS) filters are processed to US EPA Standard Method 2540 C, D and E, washed using reagent grade water, dried, desiccated and weighed to 4 decimal place resolution.
Packaging: Box of 100 filters.
Frequently Bought Together