LabExact® LabExact® Bench Protector Papers | Avantor
LabExact® LabExact® Bench Protector Papers
LabExact® LabExact® Bench Protector Papers
Certificates
LabExact® LabExact® Bench Protector Papers
LabExact® LabExact® Bench Protector Papers
2 Options Available Below
Product Details & Documents
LabExact® disposable, highly absorbent pure cellulose paper with polyethylene backing protects laboratory surfaces against liquid spills and leakage through to the bench top.
  • Lays flat with less wrinkles for sturdy glassware use
  • Tear resistant material, whether wet or dry
  • The polyethylene backing material is skid resistant, waterproof and chemical resistant
  • The absorbent paper quickly traps spilled or drained fluids
  • Smooth white surface is easy to write on
It is designed to protect laboratory surfaces against liquid spills and prevent leakage through to the bench top, necessary to maintain a safer work environment.
Certifications: Meets OSHA regulation 29CFR 1910.1030 for occupational exposure to bloodborne pathogens.
Product Family Options
Informations sur le produit
  • Désignation
    Type
    Color
    Matière
    Dimensions
    Availability
    Price
  • LabExact® bench protector paper, roll
    Smooth
    Bright white
    Cellulose/polyethylene
    48 cm ×50 m (18.9" × 164')
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $901.25
    $901.25
    Pack of 1
       $901.25
    Supplier Number: LEBPR4650M
  • Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $633.27
    $633.27
    Pack of 50
       $633.27
    Supplier Number: LEBPS4657