LabExact® PTFE Syringe Filters, Non Sterile
Fournisseur: I.W. TREMONT CO., INC.
New Product
Certificats
À propos de cet article
LabExact® disposable non sterile PTFE hydrophobic filtration media syringe filters are inherently hydrophobic; both hydrophobic and hydrophilic syringe filters are ideal for the filtration of nonpolar liquids and gases and are very resistant to solvents as well as acids or bases.
- Excellent chemical compatibility for filtering harsh chemicals that destroy other membrane materials
- Use for filtering and de-gassing chromatography solvents or filtering viscous organic-based HPLC samples
- Ideal for aerosol sampling and can be used as a moisture barrier for venting applications and vacuum pump line protection
- After pre-wetting with alcohol, may be used for fine particle removal of aqueous and mild organic solutions in Ion Chromatography and made more hydrophilic by flushing with alcohol, followed by water
- Excellent chemical compatibility for filtering harsh chemicals that destroy other membrane materials
- Use for filtering and de-gassing chromatography solvents or filtering viscous organic-based HPLC samples and ideal for aerosol sampling
All syringes feature Luer-Lok™ and pressure fitting and are ultrasonically bonded for reliability. Filter media type is clearly inscribed on barrel. Excellent unit-to-unit and lot-to-lot reproducibility and consistency. The concentric funnel shaped barrel increases volume and decreases back pressure while permitting optimal utilization of small sample volumes.
Frequently Bought Together