LabExact® Nylon Syringe Filters, Non Sterile
New Product
Nylon syringe filters are recommended for filtration of aqueous or organic/aqueous medium polar liquids and commonly used for general laboratory filtration and filtration of HPLC and GC samples prior to injection. The non-sterile disposable syringe filters with 4 mm diameter are ideal for processing small samples.
- LabExact® Disposable non-sterile Nylon/Polyamide filtration media syringe filters feature Luer-Lok™ and pressure fitting
- Hydrophilic material recommended for filtration of aqueous or organic/aqueous medium polar liquids
- Suitable for filtration of aqueous and organic-aqueous polar medium liquids and dissolution sample analysis and for use with high pH samples
They are ultrasonically bonded for reliability and the filter media type is clearly inscribed on barrel. Excellent unit-to-unit and lot-to-lot reproducibility and consistency. The concentric funnel shaped barrel increases volume and decreases back pressure.
: Since it binds proteins it is not suitable for protein recovery applications.
Frequently Bought Together