Magnetic Putty, with Magnet
Catalogue # 470355-406
Fournisseur: American Scientific
Spécifications
- Description:Magnetic putty, with neodymium magnet
- Size:91 g (3.21 oz.)
- Cat. no.:470355-406
Spécifications
À propos de cet article
Visualize magnetic force or discuss advanced materials.
- Comes with a teacher’s guide
- Putty comes with a neodymium cube magnet in storage tin
Made with micron-sized, iron-based particles distributed throughout, the astonishing, moldable Magnetic Thinking Putty takes on the properties of a magnet itself when placed in close contact with the included magnet. It’s perfect for science-loving kids, or for any adult that loves intelligent play! Use the magnet to do the 'snake charmer' trick. Or leave the magnet on your ball of putty and watch it completely engulf the magnet in minutes! What can you do with it? The possibilities are endless!