Pendulum Wave Demo | Avantor
Pendulum Wave Demo
Pendulum wave demo
Catalog # 470355-428
Supplier:   American Scientific
Fournisseur #:  3645-00
Pendulum Wave Demo
Catalog # 470355-428
Pendulum Wave Demo
Pendulum Wave Demo
Pendulum wave demo
Price
$211.56
Log in pour la disponibilité
Each
Product Details & Documents
Discuss the dependence of a pendulum's period on its length with this beautiful wave demo.
  • Teacher's guide included
  • Easily assembled in minutes
This item makes a captivating demonstration for all ages! The Pendulum Wave Demo simulates phases, beats, and frequencies. It features a stand with nine (9) 22 mm (0.88") steel ball pendulums and comes with an initiator stick, which helps to simultaneously release all pendulums. When released, the pendulums will swing back and forth to quickly form a wave pattern and then cycle through different phases. This product is great for a classroom, as a gift, or for display.
Spécifications
  • Désignation:
    Pendulum wave demo
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.