Pressure Sensor Transmitters | Avantor
Pressure Sensor Transmitters
Supplier:   PENDOTECH
Pressure Sensor Transmitters
Supplier:  PENDOTECH
Certificates
Pressure Sensor Transmitters
2 Options Available Below
Product Details & Documents
The enhanced PendoTECH® pressure sensor transmitter connects seamlessly to the PendoTECH pressure sensor, delivering a 4 to 20 mA signal linear with pressure. It maintains its convenient DIN rail mounting design for easy installation and integration into bioprocess systems.
  • The PTR features an alarm output function with a dry contact relay output
  • The PTR supports the digital communication protocol of modbus TCP
  • It has a convenient DIN rail mounting design for easy integration
Two model options are available to deliver optimal performance based on pressure range. For lowest-pressure applications, the PTR 7.5 psi (MFLX19406-66) has a default 4 to 20 mA range of –11.5 to +7.5 psi. For higher pressure applications, the PTR 75 psi (MFLX19406-67) supports the full pressure range –11.5 to +75 psi.
Ordering information: Selecting the narrowest range that meets the application requirements achieves better accuracy and repeatability. See data sheet for additional details.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • MFLX19406-66
    Pressure sensor transmitter, 7.5 psi
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,130.00
    $2,130.00
    Each
       $2,130.00
    Supplier Number: 30827316
  • MFLX19406-67
    Pressure sensor transmitter, 75 psi
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,200.00
    $2,200.00
    Each
       $2,200.00
    Supplier Number: 30827317