ContactUsLinkComponent
Pierce™ Ubiquitin Enrichment Kit, Thermo Scientific
Catalog # CAPI89899
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ Ubiquitin Enrichment Kit, Thermo Scientific
Catalog # CAPI89899
Supplier:  Thermo Scientific
Supplier Number:  89899
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ubiquitin Enrichment Kit
  • Cat. no.:
    CAPI89899
  • Supplier No.:
    89899
Specifications
About this item
The Thermo Scientific Pierce Ubiquitin Enrichment Kit facilitates the isolation of polyubiquitin protein conjugates from cultured cells and tissue samples.
  • Fast—less than 45 minutes hands-on time for binding and recovery of ubiquitinated proteins
  • Complete—includes all reagents needed for ubiquitin-modified protein enrichment from cultured cells and tissue samples
  • Flexible—sample incubation from 2 hours to overnight allows assay to be completed in several hours or in less than 30 minutes after overnight incubation
  • Robust—compatible with all Thermo Scientific Pierce Cell Lysis Products and standard RIPA formulations
  • Multiple-sample format—easily processes 1 to 15 samples concurrently
The enriched fraction can subsequently be analyzed be quantitatively or qualitatively, depending on the application. The spin-column protocol requires less than 45 minutes of hands-on prep time and incubation can range from two hours to overnight for flexibility in scheduling assay times. The components in the kit are compatible with all Poppers cell lysis products and standard RIPA formulations. It will process up to 15 samples concurrently.
Delivery: The kit contains Polyubiquitin Positive control (1000X, 50 µL), Anti-Ubiquitin Antibody (Rabbit antiserum, 5 µL), Polyubiquitin affinity resin slurry (300 µL), TBS pack (makes 500 mL), and Spin Columns (18) and Accessories.