VWR® Atlas Data Column Inserts
VWR® Atlas Data Column Inserts
Supplier:   VWR International
VWR® Atlas Data Column Inserts
Supplier:  VWR International
Certificates
VWR® Atlas Data Column Inserts
2 Options Available Below
Product Details & Documents
Data column inserts are used when data connections are required to be flush-mounted into column uprights.
  • Easy to install and remove
  • Removable black inserts for access from the front
Product Family Options
Product Information
  • Désignation
    Color
    Largeur
    Hauteur
    Availability
    Price
  • One RJ45-CAT5 Outlet
    Light neutral
    3⅝"
    24"
    Log in for availability
    Log in for availability
    Each
    Each - $236.40
    $236.40
    Each
       $236.40
  • Four RJ45-CAT5 Outlets
    Light neutral
    3⅝"
    24"
    Log in for availability
    Log in for availability
    Each
    Each - $452.28
    $452.28
    Each
       $452.28