VWR® Self-Contained Nitrogen Generator
VWR® Self-Contained Nitrogen Generator
Supplier:   Parker Hannifin
VWR® Self-Contained Nitrogen Generator
Supplier:  Parker Hannifin
Certificates
VWR® Self-Contained Nitrogen Generator
1 Options Available Below
Product Details & Documents
Self-contained membrane nitrogen generator produces LC/MS grade nitrogen with a maximum output pressure of 116psig and maximum flow capacity of 30L/min.
  • Continuous or on-demand nitrogen supply
  • Reliable, durable generator
  • Special sound insulation design ensures quiet operation
The membrane will not suppress corona needle discharge, unlike PSA and Hosmer technologies
Generator can deliver a continuous or on-demand supply of pure, phthalate-free nitrogen, making it an excellent alternative to cylinders or dewars. Reliable, durable generator has been engineered with oil-free compressors, carefully matched filtration, and membrane separation and compression technologies.
Ambient air is filtered using an inlet suction breather filter. The purified air is delivered to a long-life low pressure air compressor, which provides an air stream to hollow fiber membranes. These membranes separate the clean air into a concentrated nitrogen retentate and oxygen-enriched permeate. Prior to exiting the system, pure nitrogen retentate is delivered to a nitrogen amplification compressor to assure proper pressure, flow, and purity to the LC/MS system.
Additional applications include nebulizer gases, chemical and solvent evaporation, instrument supply and purge, evaporative light scattering equipment, and sparging. Also ideal for derivatives of ESi and APCi modes.
Specifications for Product Family
Specifications
  • Catalog No:
  • CA62700-326
  • 97021-300
  • Dimensions:
  • 70.1H x 31W x 90D cm (271/2 x 121/4 x 357/16")
  • 70.1H x 31W x 90D cm (271/2 x 121/4 x 357/16")
  • Weight:
  • 92.5 kg (204 lbs.)
  • 92.5 kg (204 lbs.)
Specifications
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • VWR® Self-Contained Nitrogen Generator
    Log in for availability
    Log in for availability
    Each
    Each - $49,290.00
    $49,290.00
    Each
       $49,290.00
    Minimum order quantity is 1
    Specifications:
    More
    Specifications
    Cat. No.
    CA62700-326
    Dimensions
    70.1H x 31W x 90D cm (271/2 x 121/4 x 357/16")
    Weight
    92.5 kg (204 lbs.)
  • VWR® Self-Contained Nitrogen Generator
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    97021-300
    Dimensions
    70.1H x 31W x 90D cm (271/2 x 121/4 x 357/16")
    Weight
    92.5 kg (204 lbs.)