Vector, E. coli BL21, Cytiva
Vector, E. coli BL21, Cytiva
E. coli BL21
Catalog # CA95017-592L
Supplier:   Cytiva
Supplier #:  27-1542-01
Vector, E. coli BL21, Cytiva
Catalog # CA95017-592L
Supplier:  Cytiva
CAS Number:  
Vector, E. coli BL21, Cytiva
Vector, E. coli BL21, Cytiva
E. coli BL21
CAS Number:  
Price
$775.62
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The E. coli BL21 strain is protease-deficient for optimal expression of a variety of recombinant proteins.
  • High level expression of a variety of recombinant proteins
  • Defective in E. coli strain B and wild-type restriction systems to permit high efficiency transformation with unmodified plasmid DNA
  • Verified by RAPD fingerprint analysis using ready-to-go RAPD analysis kit
  • Supplied lyophilised
Well suited for use with the GST gene fusion system, E. coli BL21 is an exemplary host for the expression of recombinant proteins. As an E. coli B strain, it further lacks the major protease, encoded by the lon gene, catalysing the endoproteolytic cleavage of damaged and recombinant proteins in the cell.
The protease-minus nature of BL21 makes it useful as an expression host. Since BL21 does not transform well, use an alternate strain for cloning and maintenance of the vector.
Genotype: The BL21 strain is derived from E. coli B. The genotype is F-, ompT, hsdS (rB -, mB -), gal, dcm.
Growth conditions: Resuspend lyophilised cultures in 1 ml of LB medium. Grow overnight at 37 °C before plating onto LB agar plates.
Storage conditions: −20 °C. Mix equal volumes of stationary phase culture (grown in LB medium) and glycerol. Store at –70 °C. Revive frozen glycerol stocks of BL21 by streaking onto LB agar plates.
Spécifications
Spécifications
  • Désignation:
    <i>E. coli</i> BL21