Penetration Needle, Koehler
Penetration Needle
Catalog # 52934-662
Supplier: Koehler
: K20500-00000
Jump to:
Product Details & Documents
For testing of bituminous materials
Made from hardened stainless steel with a brass shank.
: Meets ASTM D5 specifications.
Spécifications
- Désignation:Penetration Needle