Select-D and NuClean™ Spin Columns, IBI Scientific
Select-D Columns are ideal for desalting and removing unincorporated radiolabeled deoxynucleotide triphosphates from end labels, fill in, nick-translation, or random primed DNA labeling reactions
The complete operation takes a total of ten minutes. Columns come fully hydrated and ready to use. All Select-D columns are RNase and DNase free.
- Method / Format:Column (silica-based)
- Target molecule:DNA
Frequently Bought Together