VWR® Contour™ Wall Mounted Storage Cabinets with Swinging Panel Doors
Fournisseur: VWR International
Certificats
À propos de cet article
Heavy-gauge steel full-frame construction.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Wall cabinets come with two adjustable shelves.
Frequently Bought Together