Ward's® PCR Bundle
Product Details & Documents
Experience seamless PCR sampling, preparation, and storage with this specially curated bundle.
- The PCR Bundle provides educators with everything they need to facilitate precise, temperature-sensitive experiments
- Makes PCR techniques more accessible and efficient for classroom labs
Sprout® Plus mini centrifuge (470337-864) - This compact centrifuge offers user-friendly features such as tool-free snap spin rotors, quick-spin options, a braking function for quick stops, and a weighted rubberized base for stability and noise reduction.
Mini vortexer (470316-814) - Small footprint vortexer offering maximum vortex shaking for a variety of tube sizes. A simple touch start operation allows for immediate sample mixing with one hand and automatic stop when tube is lifted.
PCR cooler (76343-418) - Ice-free cooling system for PCR prep and storage. Efficiently cools PCR tubes, strips, and plates, maintaining 0 °C for over an hour and below 5 °C for up to three hours. Color change alerts when temperatures exceed 6 °C.
Low temp PCR box (76045-408) - Featuring a rotating lid that doubles as a stand. Translucent for quick cooling and sample ID, with ergonomic grips and a sturdy ABS base. Available in various colors for lab coding.
Ward’s® silicone lab mat (470316-832) - This FDA food-grade silicone lab mat is ideal for high-sterility environments, protecting counter tops from temperatures up to 200 °C (392 °F) and resisting moderate oxidizing chemicals.
Spécifications
- Désignation:PCR bundle