Ward's® Colony Counter Bundle | Avantor
Ward's® Colony Counter Bundle
Colony counter bundle
Catalog # 470357-322
Supplier:   Heathrow Scientific
Fournisseur #:  121009
Ward's® Colony Counter Bundle
Catalog # 470357-322
Ward's® Colony Counter Bundle
Ward's® Colony Counter Bundle
Colony counter bundle
Price
$695.20
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Quickly identify and tally colonies with precision using this all-inclusive package designed to streamline all colony counting procedures.
  • This colony counting bundle provides all the essential tools to streamline and enhance the accuracy of bacterial and fungal colony counting
  • Makes it easier for educators to bring hands-on microbiology lessons to life in the classroom
eCount colony counter (470311-036) - Combines an electronic counter with a Sharpie™ pen to prevent counting errors. Features count verification, dual modes, a display for up to 32 counts, and an option to disable the counter for notes.
Ward’s® mini magnifier (470311-198) - The Mini-Magnifier enhances object visibility by providing 1.75× magnification. It features a swiveling, molded lens for comfortable observation.
Ward’s® lightbox (470311-200) - The lightbox serves as a LED light source for efficient bacterial colony counting. It features a spacious viewing area (125×100 mm) suitable for various samples, and is powered by six 1.5 V AAA batteries (included).
Ward’s® silicone lab mat (470316-832) - This FDA food-grade silicone lab mat is ideal for high-sterility environments, protecting counter tops from temperatures up to 200 °C (392 °F) and resisting moderate oxidizing chemicals.
Spécifications
  • Désignation:
    Colony counter bundle