ContactUsLinkComponent
Sulfinert® Sample Loop, 5 µl, Restek
Catalog # RK22840
Supplier:  Restek
undefined
Sulfinert® Sample Loop, 5 µl, Restek
Catalog # RK22840
Supplier:  Restek
Supplier Number:  22840
Specifications
  • Capacity:
    5 µl
  • Cat. no.:
    RK22840
  • Supplier No.:
    22840
Specifications
About this item
Ideal for samples containing low concentrations of sulfur or other active compounds.
  • Sample loop sizes from 5 μL to 5 mL
  • Sulfinert®-treated
Sulfinert® treatment eliminates active sites in the loop for better active compound recovery. Sample loop features 1/16" fittings for W-type valves.