PerpetuLab Plain Apron Tables
À propos de cet article
Legs and aprons are constructed of solid red oak with heavy-duty 13-gauge steel corner braces.
- 2¼" solid square legs
- 1" epoxy resin or ¾" phenolic resin worksurface
- Non-skid adjustable glide for precise leveling
- Chemical resistant UV finish
- Available in sitting height (30") or standing height (36")
Delivered knocked down with tops attached and easy assembly of legs.
Certifications: Diversified Woodcrafts, Inc is SEFA compliant and MAS Certified Green.
14 Options Available Below
Product Details & Documents
Legs and aprons are constructed of solid red oak with heavy-duty 13-gauge steel corner braces.
- 2¼" solid square legs
- 1" epoxy resin or ¾" phenolic resin worksurface
- Non-skid adjustable glide for precise leveling
- Chemical resistant UV finish
- Available in sitting height (30") or standing height (36")
Delivered knocked down with tops attached and easy assembly of legs.
Certifications: Diversified Woodcrafts, Inc is SEFA compliant and MAS Certified Green.
Specifications for Product Family
Spécifications
- Catalog No:
- 470117-830
- 470117-834
- 470117-838
- 470117-842
- 470205-340
- 470205-346
- 470205-352
- 470117-828
- 470117-832
- 470117-836
- 470205-334
- 470205-408
- 470205-414
- 470205-426
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
Spécifications
Product Family Options
Sitting height
Informations sur le produit
- DésignationWorksurface MaterialWorksurface ThicknessW×D×HAvailabilityPrice
- Supplier Number: P7104K30NSpécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.470117-830Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationMAS Certified GreenSafer Human and Environmental Health - Supplier Number: P7204K30NSpécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.470117-834Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationMAS Certified GreenSafer Human and Environmental Health - Supplier Number: P7604K30NSpécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.470117-838Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationMAS Certified GreenSafer Human and Environmental Health - Supplier Number: P7304K30NSpécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.470117-842Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationMAS Certified GreenSafer Human and Environmental Health - Supplier Number: P7206K30NSpécifications:
More
Specifications
Cat. No.470205-340Environmentally PreferableEnvironmental certification - Supplier Number: P7306Spécifications:
More
Specifications
Cat. No.470205-346Environmentally PreferableEnvironmental certification - Supplier Number: P7606Spécifications:
More
Specifications
Cat. No.470205-352Environmentally PreferableEnvironmental certification
Standing height
Informations sur le produit
- DésignationWorksurface MaterialWorksurface ThicknessW×D×HAvailabilityPrice
- Supplier Number: P7104K36NSpécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.470117-828Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationMAS Certified GreenSafer Human and Environmental Health - Supplier Number: P7204K36NSpécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.470117-832Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationMAS Certified GreenSafer Human and Environmental Health - Supplier Number: P7604K36NSpécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.470117-836Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationMAS Certified GreenSafer Human and Environmental Health - Supplier Number: P7106Spécifications:
More
Specifications
Cat. No.470205-334Environmentally PreferableEnvironmental certification - Supplier Number: P7106K36NSpécifications:
More
Specifications
Cat. No.470205-408Environmentally PreferableEnvironmental certification - Supplier Number: P7206K36NSpécifications:
More
Specifications
Cat. No.470205-414Environmentally PreferableEnvironmental certification - Supplier Number: P7606K36NSpécifications:
More
Specifications
Cat. No.470205-426Environmentally PreferableEnvironmental certification