Drum Storage Cabinets, Eagle Manufacturing
Drum Storage Cabinets, Eagle Manufacturing
Supplier:   Eagle Manufacturing
Drum Storage Cabinets, Eagle Manufacturing
Certificates
Drum Storage Cabinets, Eagle Manufacturing
Drum Storage Cabinets, Eagle Manufacturing
4 Options Available Below
Product Details & Documents
Perfect for safe storage of flammable liquid drums near work areas.
  • Finish is caution-yellow enamel with red warning
  • Complete with grounding attachments, three-point key lock, and 5 cm (2") raised leakproof door sill
  • Drum cabinet ramp attaches firmly to cabinet sill and accommodates most dollies for simplified loading and unloading
  • Ramp is constructed of heavy duty galvanized steel, and can withstand loads up to 680 kg (1500 lbs.)
Horizontal storage cabinet interior is equipped with drum cradle guide rails. Rails are 13.9 cm (5½") wide and are set 27.9 cm (11") apart to accommodate dollies with wheels 27.9 cm (11") to 55.8 cm (22") apart. Vertical storage cabinet is equipped with adjustable drum supports.
Safely store 208.2 L (55 gal.) drums in a horizontal or vertical position in these two-door, 18-gauge steel cabinets. Double-walled sides, top, bottom, and doors have 3.8 cm (1½") air between walls. Both vents, with 5 cm (2") pipe thread fittings, have fire baffle and cap.
Certifications: Meets OSHA and NFPA Code 30 requirements.
Product Family Options
Product Information
  • Désignation
    Type
    Longueur
    Profondeur
    Hauteur
    Weight
    Availability
    Price
  • Cabinet for horizontal drum
    Manual door
    20"
    20"
    28"
    385 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $6,228.58
    $6,228.58
    Each
       $6,228.58
    Supplier Number: 1928X
  • Cabinet for vertical drum
    Manual door
    20 x 31 x 36⅝"
    334 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $4,643.60
    $4,643.60
    Each
       $4,643.60
    Supplier Number: 1926X
  • Cabinet for vertical drum
    Self-closing door
    15¾ x 16¾ x 21¼"
    334 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $5,225.91
    $5,225.91
    Each
       $5,225.91
    Supplier Number: 2610X
  • Cabinet for horizontal drum
    Self-closing door
    18 x 43 x 44⅝"
    385 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $6,997.87
    $6,997.87
    Each
       $6,997.87
    Minimum order quantity is 1
    Supplier Number: 2810X
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Overpack dolly
    Overpack dolly
    56610-165
    Log in for availability
    Each - $2,684.99
Product Information
  • Catalog #
    Availability
    Price
  • Drum lifter
    Drum lifter
    56610-167
    Log in for availability
    Each - $887.14