About this item
Purpose-built for life science applications, FARRAR®’s ULC series including the Ultra-Low Chamber (ULC-190), Ultra-Low Chamber (ULC-328), and the new Ultra-Low Chamber (ULC-259) are the only −20 to −80 °C, forced-air, ultra-low-temperature freezers on the market with air and water-cooled options.
- Optimized footprint - 50% less floorspace versus standard ULT configurations with six times the capacity (one ULC equals six ULTs)
- Industry's fastest temperature recovery - 10× faster temperature recovery from door opening helps ensure product integrity
- Integrated, fully redundant systems (control, refrigerations, airflow) - Eliminates need for backup freezer
- Automatic defrost - Never worry about taking units out of service to manually defrost
- Extremely tight temperature uniformity at any setpoint - At +4, −80 °C or any temperature in between, count on better than ±3 °C anywhere inside the ULC, even during a defrost cycle
- Engineered with material handling in mind - Optional racking, shelving, drawers and carts optimize process and workflow (available for all models)
- Equipment performance/temperature monitoring (MODbus) - Enables easy connection to nearly every building management and automation system (BAS/BMS)
Forced air convection: An industry game changer - FARRAR® pioneered forced air convection technology in its reach-in chambers. This key differentiator affects uniformity. The chambers uniformly force air, at the targeted temperature, throughout the chamber. This means FARRAR® freezer and freeze/thaw equipment is up to 10 times more efficient at removing heat than the traditional cold-wall heat displacement process. Recovery from door opening is critical to helping ensure product integrity. FARRAR® ULC Series chambers dual refrigeration units recover to −80 °C in less than 25 minutes from a 1 minute door opening, versus several hours experienced with competitor solutions.
ULC-190 Standard Set-up, Shelves
* ULC 190 Chamber - #6700617
* Refrigeration System - #6700028 (air-cooled), #6000442 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* Standard ULC-190 Shelf Kit - #6700645
ULC-259 Standard Set-up, Shelves
* ULC 259 Chamber - #6700619
* Refrigeration System - #6700028 (air-cooled), #600029 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-259 Standard Shelf System - #6700775
ULC-259 Standard Set-up, Drawers
* ULC 259 Chamber - #6700619
* Refrigeration System - #6700028 (air-cooled), #600029 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-259 Drawer System - #6700761
ULC-259 Standard Set-up, Biobank
* ULC 259 Chamber - #6700619
* Refrigeration System - #6700028 (air-cooled), #600029 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-259 Biorack Assembly - #6700769
ULC-328 Standard Set-up, Pallet Rack
* ULC 328 Chamber - #6700615
* Refrigeration System - #6700028 (air-cooled), #600029 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-328 Pallet Rack Assembly - #6700401
Specifications
- Catalog No:
- 77592-006
- 77591-996
- 77592-008
- 77592-010
- 77592-000
- 77592-012
- 77591-998
- 77592-014
- 77592-004
- 77592-002
- Cat. No.:
- 77592-006
- 77591-996
- 77592-008
- 77592-010
- 77592-000
- 77592-012
- 77591-998
- 77592-014
- 77592-004
- 77592-002
- Air circulation:
- Forced air circulation
- Forced air circulation
- Forced air circulation
- Forced air circulation
- Forced air circulation
- Forced air circulation
- Forced air circulation
- Forced air circulation
- Forced air circulation
- Forced air circulation
- Display:
- HMI touchscreen provides visual temperature profile progress with export capability
- HMI touchscreen provides visual temperature profile progress with export capability
- HMI touchscreen provides visual temperature profile progress with export capability
- HMI touchscreen provides visual temperature profile progress with export capability
- HMI touchscreen provides visual temperature profile progress with export capability
- HMI touchscreen provides visual temperature profile progress with export capability
- HMI touchscreen provides visual temperature profile progress with export capability
- HMI touchscreen provides visual temperature profile progress with export capability
- HMI touchscreen provides visual temperature profile progress with export capability
- HMI touchscreen provides visual temperature profile progress with export capability
- Defrost:
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Electrical:
- 208-230VAC, Single Phase, 60Hz
- 208-230VAC, Single Phase, 60Hz
- 208-230VAC, Single Phase, 60Hz
- 208-230VAC, Single Phase, 60Hz
- 208-230VAC, Single Phase, 60Hz
- 208-230VAC, Single Phase, 60Hz
- 208-230VAC, Single Phase, 60Hz
- 208-230VAC, Single Phase, 60Hz
- 208-230VAC, Single Phase, 60Hz
- 208-230VAC, Single Phase, 60Hz
- Shelves/drawers:
- Includes ULC-259 Drawer System which provides 6 drawers on each side of unit. Each drawer has 4 individual pans.
- Includes ULC-190 Standard Shelf Kit (3 shelves behind each door)
- Includes ULC-259 Standard Shelf System (3 shelves per unit)
- Includes ULC-259 Standard Shelf System (3 shelves per unit)
- Includes ULC-259 Biorack Assembly. Designed to hold 112 of our 572 Cryoracks (accessory).
- Includes ULC-328 Pallet Rack. Designed to accommodate 2 pallets for bulk storage behind each door.
- Includes ULC-190 Standard Shelf Kit (3 shelves behind each door)
- Includes ULC-328 Pallet Rack. Designed to accommodate 2 pallets for bulk storage behind each door.
- Includes ULC-259 Drawer System which provides 6 drawers on each side of unit. Each drawer has 4 individual pans.
- Includes ULC-259 Biorack Assembly. Designed to hold 112 of our 572 Cryoracks (accessory).
- Weight:
- 3,500 lbs. (1,588 kg)
- 3,150 lbs. (1,429 kg)
- 3,500 lbs. (1,588 kg)
- 3,500 lbs. (1,588 kg)
- 3,500 lbs. (1,588 kg)
- 3,900 lbs. (1,769 kg)
- 3,150 lbs. (1,429 kg)
- 3,900 lbs. (1,769 kg)
- 3,500 lbs. (1,588 kg)
- 3,500 lbs. (1,588 kg)
- Shipping weight:
- 4,100 lbs. (1,860 kg)
- 3,900 lbs. (1,769 kg)
- 4,100 lbs. (1,860 kg)
- 4,100 lbs. (1,860 kg)
- 4,100 lbs. (1,860 kg)
- 4,500 lbs. (2,132 kg)
- 3,900 lbs. (1,769 kg)
- 4,500 lbs. (2,132 kg)
- 4,100 lbs. (1,860 kg)
- 4,100 lbs. (1,860 kg)