VWR® Large-Volume, Heavy-Walled Bulbs
Fournisseur: VWR International
Certificats
À propos de cet article
Easy-to-grip bulbs feature heavy-walled necks for use with pipettes and syringes.
- Heavy-walled design
- Natural rubber fabrication
- Easy to grip
Made of natural red rubber. The 150 ml bulb (82024-572) features reinforcing ribs to aid in larger volume applications.
Frequently Bought Together