Electric Hot Plate Set, Carver
53881-007
: Carver
- Cat. no.:53881-007
- Electrical:115 V
- Supplier No.:2101
For use with standard laboratory presses (see 53880-048 series)
This hot plate set incorporates a top and bottom aluminum hot plate. These 15.2cm (6") plates have individual temperature controllers and thermometers. They provide heat from 65.6° to 260°C (150° to 500°F).