LabExact® CFP542 Quantitative Ashless Hardened Cellulose Filter Papers, 2 to 3 µm
Supplier: I.W. TREMONT CO., INC.
New Product
Certificates
About this item
Grade CFP542 quantitative cellulose filter media is ashless and hardened with increased wet strength.
- Very slow flow rates and retention of approximately 2 to 3 µm
Packaging: Box of 100 filters.
Specifications
- Catalog No:
- 77590-758
- 77590-756
- 77590-754
- 77590-752
- 77590-750
- 77590-748
- Ash content:
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- Basis weight:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Retention:
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm