LabExact® CFP541 Quantitative Ashless Hardened Cellulose Filter Papers, 21 to 23 µm
New Product
Grade CFP541 quantitative cellulose filter media is ashless and hardened with increased wet strength.
- Fast flow rates and retention of approximately 21 to 23 µm
: Box of 100 filters.
- Catalog No:
- 77590-736
- 77590-746
- 77590-744
- 77590-742
- 77590-740
- 77590-738
- Ash content:
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- Basis weight:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Retention:
- 21 - 23 µm
- 21 - 23 µm
- 21 - 23 µm
- 21 - 23 µm
- 21 - 23 µm
- 21 - 23 µm
Frequently Bought Together