LabExact® CFP540 Quantitative Ashless Hardened Cellulose Filter Paper, 7 - 8 µm
Supplier: I.W. TREMONT CO., INC.
New Product
Certificates
About this item
Grade CFP540 quantitative cellulose filter media is ashless and hardened with increased wet strength.
- Medium flow rates and retention of approximately 7 to 8 µm
Extremely low ash content of <0.01%.
Packaging: Box of 100 filters.
Specifications
- Catalog No:
- 77590-734
- 77590-732
- 77590-730
- 77590-728
- 77590-726
- 77590-724
- Ash content:
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- Basis weight:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Retention:
- 7 - 8 µm
- 7 - 8 µm
- 7 - 8 µm
- 7 - 8 µm
- 7 - 8 µm
- 7 - 8 µm