LabExact® CFP44 Quantitative Ashless Cellulose Filter Papers, 3 µm
Supplier: I.W. TREMONT CO., INC.
New Product
Certificates
About this item
Very similar to CFP42 but demonstrating a slightly wider retention rate within a similar flow rate.
Typically used in the analysis of samples requiring separation of very fine precipitates.
Packaging: Box of 100 filters.
Specifications
- Catalog No:
- 77590-694
- 77590-692
- 77590-690
- 77590-688
- 77590-698
- 77590-696
- Ash content:
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- Basis weight:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Retention:
- 3.0 µm
- 3.0 µm
- 3.0 µm
- 3.0 µm
- 3.0 µm
- 3.0 µm