LabExact® CFP43 Quantitative Ashless Cellulose Filter Papers, 15 to 17 µm
Supplier: I.W. TREMONT CO., INC.
New Product
Certificates
About this item
Positioned as an intermediate within the quantitative cellulose family, this grade demonstrates medium retention and flow rates.
Ideal for gravimetric analysis of soil samples, surface water testing procedures, and use in air sample monitoring equipment.
Packaging: Box of 100 filters.
Specifications
- Catalog No:
- 77590-684
- 77590-682
- 77590-680
- 77590-678
- 77590-686
- 77590-676
- 77590-674
- 77590-672
- Ash content:
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- Basis weight:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Retention:
- 15 - 17 µm
- 15 - 17 µm
- 15 - 17 µm
- 15 - 17 µm
- 15 - 17 µm
- 15 - 17 µm
- 15 - 17 µm
- 15 - 17 µm