LabExact® CFP42 Quantitative Ashless Cellulose Filter Papers, 2.5 µm
Supplier: I.W. TREMONT CO., INC.
New Product
Certificates
About this item
This media demonstrates an extremely high retention rate for a cellulose filter media.
- Ideal for any gravimetric analysis of very fine precipitates
The uniquely high retention rate make it ideal for any gravimetric analysis of very fine precipitates.
Packaging: Box of 100 filters.
Specifications
- Catalog No:
- 77590-660
- 77590-658
- 77590-656
- 77590-670
- 77590-668
- 77590-664
- 77590-666
- 77590-662
- Ash content:
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- Basis weight:
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- Retention:
- 2.5 µm
- 2.5 µm
- 2.5 µm
- 2.5 µm
- 2.5 µm
- 2.5 µm
- 2.5 µm
- 2.5 µm