LabExact® CFP41 Quantitative Ashless Cellulose Filter Papers, 20 µm
Fournisseur: I.W. TREMONT CO., INC.
New Product
Certificats
À propos de cet article
Very fast flow rates and loose retention characteristics make this grade ideal for initial separation of gelatinous precipitates.
Packaging: Box of 100 filters.
Spécifications
- Catalog No:
- 77590-632
- 77590-644
- 77590-642
- 77590-648
- 77590-646
- 77590-638
- 77590-636
- 77590-634
- Ash content:
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- Basis weight:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Retention:
- 20 µm
- 20 µm
- 20 µm
- 20 µm
- 20 µm
- 20 µm
- 20 µm
- 20 µm