LabExact® CFP40 Quantitative Ashless Cellulose Filter Papers, 8 µm
Supplier: I.W. TREMONT CO., INC.
New Product
Certificates
About this item
Demonstrating medium retention and flow rates, this media is widely used in many general quantitative procedures in both liquid and gas.
- Medium retention and flow rates
Recommended procedures include, standard environmental test procedures such as soil sample analysis and the collection of trace elements and radionuclides in air samples. Also used in liquid food tests for determination of sediments, primary analysis of cements and slurries, and sample preparation prior to spectrophotometry.
Packaging: Box of 100 filters.
Specifications
- Catalog No:
- 77590-614
- 77590-612
- 77590-610
- 77590-608
- 77590-622
- 77590-620
- 77590-618
- 77590-616
- Ash content:
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- <0.01%
- Basis weight:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Retention:
- 8 µm
- 8 µm
- 8 µm
- 8 µm
- 8 µm
- 8 µm
- 8 µm
- 8 µm