LabExact® CFP3 Qualitative Cellulose Filter Papers, 6 µm
Supplier: I.W. TREMONT CO., INC.
New Product
Certificates
About this item
Virtually identical to CFP1, but twice as thick, resulting in a significantly slower flow rate with a retention of approximately 6 µm.
This media does not clog as easily as the other qualitative cellulose types, which allows for much higher sample volume usage. This media also demonstrates very high levels of absorbency, permitting the media to be used as a sample conveyance substrate.
Packaging: Box of 100 filters.
Specifications
- Catalog No:
- 77590-604
- 77590-602
- 77590-600
- 77590-598
- 77590-606
- 77590-596
- 77590-594
- 77590-592
- Ash content:
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- Basis weight:
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- Retention:
- 6 µm
- 6 µm
- 6 µm
- 6 µm
- 6 µm
- 6 µm
- 6 µm
- 6 µm