LabExact® Extraction Thimbles, Glass Microfiber
LabExact® Extraction Thimbles, Glass Microfiber
Certificates
LabExact® Extraction Thimbles, Glass Microfiber
LabExact® Extraction Thimbles, Glass Microfiber
20 Options Available Below
Product Details & Documents
LabExact® high quality glass microfiber extraction thimbles available in a variety of dimensions to fit most Soxhlet extraction units, ideal for gravimetric methods for hot environments and acidic gases as well as extraction methods common to biochemical analysis.
  • Extraction thimbles are manufactured with 100% pure borosilicate glass binderless microfibers
They have a seamless and consistent 1.5 mm wall thickness, smooth interior surface and superior mechanical strength and retention.
Common applications include gravimetric methods for hot environments and acidic gases as well as extraction methods common to biochemical analysis.
Product Family Options
Informations sur le produit
  • Matière
    Désignation
    I.D.×H
    Épaisseur de paroi
    Particle Retention
    Availability
    Price
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    16×50 mm
    1.0 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $351.99
    $351.99
    Box of 25
       $351.99
    Supplier Number: LEGT9103
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-184
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    16×100 mm
    1.0 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $351.99
    $351.99
    Box of 25
       $351.99
    Supplier Number: LEGT9104
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-186
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    22×80 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $351.99
    $351.99
    Box of 25
       $351.99
    Supplier Number: LEGT9106
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-188
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    25×90 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $351.99
    $351.99
    Box of 25
       $351.99
    Supplier Number: LEGT9107
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-190
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    25×100 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $351.99
    $351.99
    Box of 25
       $351.99
    Supplier Number: LEGT9108
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-192
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    27×80 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $351.99
    $351.99
    Box of 25
       $351.99
    Supplier Number: LEGT9109
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-194
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    28×60 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $351.99
    $351.99
    Box of 25
       $351.99
    Supplier Number: LEGT9111
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-196
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    28×120 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $413.24
    $413.24
    Box of 25
       $413.24
    Supplier Number: LEGT9114
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-198
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    30×150 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $413.24
    $413.24
    Box of 25
       $413.24
    Supplier Number: LEGT9118
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-200
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    33×80 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $351.99
    $351.99
    Box of 25
       $351.99
    Supplier Number: LEGT9120
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-202
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    33×90 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $422.36
    $422.36
    Box of 25
       $422.36
    Supplier Number: LEGT9121
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-204
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    33×94 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $422.36
    $422.36
    Box of 25
       $422.36
    Supplier Number: LEGT9122
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-206
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    33×100 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $422.36
    $422.36
    Box of 25
       $422.36
    Supplier Number: LEGT9123
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-208
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    33×118 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $505.07
    $505.07
    Box of 25
       $505.07
    Supplier Number: LEGT9124
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-210
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    33×205 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $688.73
    $688.73
    Box of 25
       $688.73
    Supplier Number: LEGT9126
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-212
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    35×150 mm
    1.5 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $596.88
    $596.88
    Box of 25
       $596.88
    Supplier Number: LEGT9128
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-214
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    43×123 mm
    2.0 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $637.27
    $637.27
    Box of 25
       $637.27
    Supplier Number: LEGT9131
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-216
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, standard format
    48×230 mm
    2.0 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $914.51
    $914.51
    Box of 25
       $914.51
    Supplier Number: LEGT9135
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-218
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, collar and recess format
    57×315 mm
    2.0 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $1,374.40
    $1,374.40
    Box of 25
       $1,374.40
    Supplier Number: LEGT9138
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-220
  • Binderless glass microfiber
    LabExact® Extraction thimbles glass microfiber, collar and recess format
    75×330 mm
    2.0 mm
    5 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $1,374.40
    $1,374.40
    Box of 25
       $1,374.40
    Supplier Number: LEGT9143
    Spécifications:
    More
    Specifications
    Cat. No.
    77591-222