LabExact® Extraction Thimbles, Cellulose
LabExact® Extraction Thimbles, Cellulose
LabExact® Extraction Thimbles, Cellulose
LabExact® Extraction Thimbles, Cellulose
LabExact® high quality cellulose extraction thimbles are available in a variety of dimensions to fit most Soxhlet extraction units.
  • Extraction thimbles are manufactured with high purity cellulose fibers and have a consistent wall thickness
  • Available as fast flow, with lid and extra density
Common applications include air and waste gas analysis, collection of solid particles such as dust, concrete slurry evaluation, analysis of pesticide residues, oil/fat content of solid foods and oil and grease analysis of solid wastes.
Product Family Options
  • Matière
    Désignation
    I.D.×H
    Épaisseur de paroi
    Particle Retention
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    8×40 mm
    1.0 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5001
    Specifications
    Cat. No.
    77590-784
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    9×50 mm
    1.0 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5002
    Specifications
    Cat. No.
    77590-786
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    15×50 mm
    1.0 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5003
    Specifications
    Cat. No.
    77590-788
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    15×100 mm
    1.0 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5004
    Specifications
    Cat. No.
    77590-790
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    20×80 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $195.05
    $195.05
    Box of 25
       $195.05
     : LECT5005
    Specifications
    Cat. No.
    77590-792
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    22×80 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $195.05
    $195.05
    Box of 25
       $195.05
     : LECT5006
    Specifications
    Cat. No.
    77590-794
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    23×90 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5007
    Specifications
    Cat. No.
    77590-796
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    23×100 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5008
    Specifications
    Cat. No.
    77590-798
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    27×80 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5009
    Specifications
    Cat. No.
    77590-800
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    27×100 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5010
    Specifications
    Cat. No.
    77590-802
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    27×60 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5011
    Specifications
    Cat. No.
    77590-804
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    28×100 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5013
    Specifications
    Cat. No.
    77590-806
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    28×120 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $209.93
    $209.93
    Box of 25
       $209.93
     : LECT5014
    Specifications
    Cat. No.
    77590-808
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    28×80 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5015
    Specifications
    Cat. No.
    77590-810
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    28×90 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5016
    Specifications
    Cat. No.
    77590-812
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    29×100 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $209.93
    $209.93
    Box of 25
       $209.93
     : LECT5017
    Specifications
    Cat. No.
    77590-814
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    30×150 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $265.48
    $265.48
    Box of 25
       $265.48
     : LECT5018
    Specifications
    Cat. No.
    77590-816
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    30×60 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5019
    Specifications
    Cat. No.
    77590-818
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    30×80 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $165.45
    $165.45
    Box of 25
       $165.45
     : LECT5020
    Specifications
    Cat. No.
    77590-820
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    30×90 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $218.63
    $218.63
    Box of 25
       $218.63
     : LECT5021
    Specifications
    Cat. No.
    77590-822
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    33×94 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $235.90
    $235.90
    Box of 25
       $235.90
     : LECT5022
    Specifications
    Cat. No.
    77590-824
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    30×100 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $221.98
    $221.98
    Box of 25
       $221.98
     : LECT5023
    Specifications
    Cat. No.
    77590-826
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    31×118 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $262.33
    $262.33
    Box of 25
       $262.33
     : LECT5024
    Specifications
    Cat. No.
    77590-828
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    31×130 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $283.05
    $283.05
    Box of 25
       $283.05
     : LECT5025
    Specifications
    Cat. No.
    77590-830
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    33×205 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $355.73
    $355.73
    Box of 25
       $355.73
     : LECT5026
    Specifications
    Cat. No.
    77590-832
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    34×120 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $262.33
    $262.33
    Box of 25
       $262.33
     : LECT5027
    Specifications
    Cat. No.
    77590-834
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    34×150 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $308.86
    $308.86
    Box of 25
       $308.86
     : LECT5028
    Specifications
    Cat. No.
    77590-836
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    38×200 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $413.86
    $413.86
    Box of 25
       $413.86
     : LECT5029
    Specifications
    Cat. No.
    77590-838
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    40×150 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $340.33
    $340.33
    Box of 25
       $340.33
     : LECT5030
    Specifications
    Cat. No.
    77590-840
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    40×123 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $306.82
    $306.82
    Box of 25
       $306.82
     : LECT5031
    Specifications
    Cat. No.
    77590-842
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    43×130 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $335.22
    $335.22
    Box of 25
       $335.22
     : LECT5032
    Specifications
    Cat. No.
    77590-844
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    48×145 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $400.86
    $400.86
    Box of 25
       $400.86
     : LECT5033
    Specifications
    Cat. No.
    77590-846
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    48×200 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $469.42
    $469.42
    Box of 25
       $469.42
     : LECT5034
    Specifications
    Cat. No.
    77590-848
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    48×230 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $472.30
    $472.30
    Box of 25
       $472.30
     : LECT5035
    Specifications
    Cat. No.
    77590-850
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    51×145 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $451.87
    $451.87
    Box of 25
       $451.87
     : LECT5036
    Specifications
    Cat. No.
    77590-852
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    51×180 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $481.05
    $481.05
    Box of 25
       $481.05
     : LECT5037
    Specifications
    Cat. No.
    77590-854
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    57×315 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $714.26
    $714.26
    Box of 25
       $714.26
     : LECT5038
    Specifications
    Cat. No.
    77590-856
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    60×180 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $472.30
    $472.30
    Box of 25
       $472.30
     : LECT5039
    Specifications
    Cat. No.
    77590-858
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    55×275 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $597.63
    $597.63
    Box of 25
       $597.63
     : LECT5040
    Specifications
    Cat. No.
    77590-860
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    75×160 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $539.33
    $539.33
    Box of 25
       $539.33
     : LECT5041
    Specifications
    Cat. No.
    77590-862
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    68×250 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $641.36
    $641.36
    Box of 25
       $641.36
     : LECT5042
    Specifications
    Cat. No.
    77590-864
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    70×330 mm
    2.0 mm
    5 µm
    Box of 25
    Box of 25 - $787.14
    $787.14
    Box of 25
       $787.14
     : LECT5043
    Specifications
    Cat. No.
    77590-866
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    33×80 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $182.82
    $182.82
    Box of 25
       $182.82
     : LECT5951
    Specifications
    Cat. No.
    77590-888
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, standard format
    90×200 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $932.75
    $932.75
    Box of 25
       $932.75
     : LECT5055
    Specifications
    Cat. No.
    77590-868
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, collar and recess format
    79×155 mm
    5 µm
    Box of 25
    Box of 25 - $787.14
    $787.14
    Box of 25
       $787.14
     : LECT5500
    Specifications
    Cat. No.
    77590-886
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, extra density format
    22×80 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5406
    Specifications
    Cat. No.
    77590-882
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, extra density format
    30×100 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $218.63
    $218.63
    Box of 25
       $218.63
     : LECT5423
    Specifications
    Cat. No.
    77590-884
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, fast flow format
    20×80 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5105
    Specifications
    Cat. No.
    77590-870
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, fast flow format
    28×90 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
     : LECT5116
    Specifications
    Cat. No.
    77590-872
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, fast flow format
    31×205 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $355.73
    $355.73
    Box of 25
       $355.73
     : LECT5126
    Specifications
    Cat. No.
    77590-874
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, with lid
    30×80 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $310.97
    $310.97
    Box of 25
       $310.97
     : LECT5220
    Specifications
    Cat. No.
    77590-876
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, with lid
    30×100 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $364.40
    $364.40
    Box of 25
       $364.40
     : LECT5223
    Specifications
    Cat. No.
    77590-878
  • Cotton linter cellulose
    LabExact® Extraction thimbles cellulose, with lid
    31×130 mm
    1.5 mm
    5 µm
    Box of 25
    Box of 25 - $428.62
    $428.62
    Box of 25
       $428.62
     : LECT5225
    Specifications
    Cat. No.
    77590-880