Strip Shields, Trax | Avantor
Strip Shields, Trax
Strip Shields, Trax
Certificates
Strip Shields, Trax
Strip Shields, Trax
5 Options Available Below
Product Details & Documents
Designed for use in applications where access to a clean enclosure is a concern, but traffic patterns are light and restricted to personnel only (no carts or equipment)
Pre-assembled strip shields are 40mil thick and comprise 20.3cm (8") wide material overlapped 5.1cm (2"). Shipping weights vary depending on size.
Ordering information: Dual-Lock™ attachment system is included with each strip shield.
Product Family Options
Informations sur le produit
  • Width
    Longueur
    Availability
    Price
  • 3 ft.
    7 ft.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $737.65
    $737.65
    Each
       $737.65
    Supplier Number: SS40ST37
  • 4 ft.
    8 ft.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $984.33
    $984.33
    Each
       $984.33
    Supplier Number: SS40ST48
  • 6 ft.
    8 ft.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,411.96
    $1,411.96
    Each
       $1,411.96
    Supplier Number: SS40ST68
  • 8 ft.
    9 ft.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,130.35
    $2,130.35
    Each
       $2,130.35
    Supplier Number: SS40ST89
  • 10 ft.
    10 ft.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,812.90
    $2,812.90
    Each
       $2,812.90
    Supplier Number: SS40ST1010