LabExact® CFP1234 Phase Separating Cellulose Filter Papers, 11 µm
À propos de cet article
These cellulose filters create a hydrophobic barrier that can replace a separatory funnel to separate organic solvents from aqueous phase.
- This grade demonstrates a fast flow rate with similar retention to Qualitative Grade CFP1 filter paper
- Paper is treated with immobilized silicone which allows solvent to pass and retain aqueous phase
- Reduces solvent volume waste in many applications and aids in greater recoveries
Widely used in environmental studies to reduce separation time significantly compared to a separatory funnel. Quadrant fold or cone fold for use in a filter funnel, or simply drop the filter disk into a Buchner funnel. May be stacked with finer porosity filter paper or used in series for final filtration. This is a mature product and lab staple written into many standard methods and SOP's.
Packaging: Box of 100 filters.
6 Options Available Below
Product Details & Documents
These cellulose filters create a hydrophobic barrier that can replace a separatory funnel to separate organic solvents from aqueous phase.
- This grade demonstrates a fast flow rate with similar retention to Qualitative Grade CFP1 filter paper
- Paper is treated with immobilized silicone which allows solvent to pass and retain aqueous phase
- Reduces solvent volume waste in many applications and aids in greater recoveries
Widely used in environmental studies to reduce separation time significantly compared to a separatory funnel. Quadrant fold or cone fold for use in a filter funnel, or simply drop the filter disk into a Buchner funnel. May be stacked with finer porosity filter paper or used in series for final filtration. This is a mature product and lab staple written into many standard methods and SOP's.
Packaging: Box of 100 filters.
Specifications for Product Family
Spécifications
- Catalog No:
- 77590-534
- 77590-536
- 77590-538
- 77590-540
- 77590-542
- 77590-544
- Weight:
- 88 g/m² basis weight
- 88 g/m² basis weight
- 88 g/m² basis weight
- 88 g/m² basis weight
- 88 g/m² basis weight
- 88 g/m² basis weight
- Thickness:
- 0.2 mm
- 0.2 mm
- 0.2 mm
- 0.2 mm
- 0.2 mm
- 0.2 mm
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Particle retention:
- 11 µm
- 11 µm
- 11 µm
- 11 µm
- 11 µm
- 11 µm
Spécifications
Product Family Options
Informations sur le produit
- DésignationØAvailabilityPrice
- Supplier Number: LECFP12340700Spécifications:
More
Specifications
Cat. No.77590-534Weight88 g/m² basis weightThickness0.2 mmFlow rateFastParticle retention11 µm - Supplier Number: LECFP12340900Spécifications:
More
Specifications
Cat. No.77590-536Weight88 g/m² basis weightThickness0.2 mmFlow rateFastParticle retention11 µm - Supplier Number: LECFP12341100Spécifications:
More
Specifications
Cat. No.77590-538Weight88 g/m² basis weightThickness0.2 mmFlow rateFastParticle retention11 µm - Supplier Number: LECFP12341250Spécifications:
More
Specifications
Cat. No.77590-540Weight88 g/m² basis weightThickness0.2 mmFlow rateFastParticle retention11 µm - Supplier Number: LECFP12341500Spécifications:
More
Specifications
Cat. No.77590-542Weight88 g/m² basis weightThickness0.2 mmFlow rateFastParticle retention11 µm - Supplier Number: LECFP12341850Spécifications:
More
Specifications
Cat. No.77590-544Weight88 g/m² basis weightThickness0.2 mmFlow rateFastParticle retention11 µm