Deck-Mounted Gooseneck Mixing Faucets, with Nylon Hooded Handles, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
À propos de cet article
Deck-mounted mixing faucets are made of white epoxy/polyester hybrid powder-coated forged brass.
- Faucet valves are self-contained renewable units with replaceable stainless steel seats
They have a serrated hose end, black or color-coded nylon hooded handles, and 1 cm (3/8") copper tubes on inlets. Faucets are available with 15.2 cm (6") or 20.3 cm (8") spread swinging gooseneck, with or without a vacuum breaker.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Frequently Bought Together