Deck-Mounted Gooseneck Mixing Faucets with Vacuum Breaker
Fournisseur: WaterSaver Faucet
Certificats
À propos de cet article
Deck-mounted mixing faucets are made of chrome-plated forged brass and provided with a serrated hose end, four-arm handles, and 1 cm (³/₈") diameter copper tubes on inlets.
- Faucet valves are self-contained renewable units with replaceable stainless steel seats
Faucets are available with 15.2 cm (6"), 20.3 cm (8"), or 30.5 cm (12") spread gooseneck, all with a vacuum breaker.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Informations sur les accessoires: Chrome-plated brass faucet accessories include an aerator and an aspirator faucet outlet.
Frequently Bought Together