VWR® Powerpette® Plus Pipette Controller
À propos de cet article
The Powerpette plus pipette controller is a compact and portable device that features multiple user-selectable modes and speeds to perform a wide range of liquid handling applications.
- Compact, self-contained design
- Designed for left-hand or right-hand use
- Suitable for a wide range of liquid handling applications
- Accommodates pipettes ranging from 1 to 100 ml
This lightweight, rechargeable pipette controller is ideal for a wide range of liquid handling applications. Controller features user-selectable aspirating and dispensing speeds and can be used in either 'to deliver' or 'blowout' mode. Compact, self-contained design with precision molding and concave finger triggers for comfortable handling. Pistol-style grip and an angled pipette holder nozzle allow use in small spaces such as laminar flow hoods or biological safety cabinets. Bench stand enables unit to be parked in upright position or with a pipette fitted. Suitable for large or small hands and for both left- and right-handed users. Works with pipettes ranging from 1 to 100 ml.
The Powerpette® Plus Pipette Controller is supplied with a 3-year material and workmanship warranty, effective from the date of delivery.
Certifications: CE marked.
Ordering information: Unit is fitted with a 0.45 µm filter. Also supplied with manufacturer’s 3-year warranty, one 0.45 µm and one 0.2 µm hydrophobic filter, charger, and bench stand.
0 Options Available Below
Product Details & Documents
The Powerpette plus pipette controller is a compact and portable device that features multiple user-selectable modes and speeds to perform a wide range of liquid handling applications.
- Compact, self-contained design
- Designed for left-hand or right-hand use
- Suitable for a wide range of liquid handling applications
- Accommodates pipettes ranging from 1 to 100 ml
This lightweight, rechargeable pipette controller is ideal for a wide range of liquid handling applications. Controller features user-selectable aspirating and dispensing speeds and can be used in either 'to deliver' or 'blowout' mode. Compact, self-contained design with precision molding and concave finger triggers for comfortable handling. Pistol-style grip and an angled pipette holder nozzle allow use in small spaces such as laminar flow hoods or biological safety cabinets. Bench stand enables unit to be parked in upright position or with a pipette fitted. Suitable for large or small hands and for both left- and right-handed users. Works with pipettes ranging from 1 to 100 ml.
The Powerpette® Plus Pipette Controller is supplied with a 3-year material and workmanship warranty, effective from the date of delivery.
Certifications: CE marked.
Ordering information: Unit is fitted with a 0.45 µm filter. Also supplied with manufacturer’s 3-year warranty, one 0.45 µm and one 0.2 µm hydrophobic filter, charger, and bench stand.