Junkin™ Safety First Aid Blanket
Catalogue # 56612-689
Fournisseur: Junkin
Small Business Enterprise
Spécifications
- Size:157.2 x 208.2 cm (62 x 82")
- Cat. no.:56612-689
- Supplier No.:JSA-502
Spécifications
À propos de cet article
For first aid/rescues.
Keeps rescue victims warm and dry. Warm utility blanket is machine washable. Made of 70% wool, 30% synthetic fibers.